Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SARS-CoV-2 NSP3 His-tag Recombinant Protein

Catalog No. PIRP88283
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIRP88283 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIRP88283 Supplier Invitrogen™ Supplier No. RP88283
Add to Cart
Add to Cart

Recombinant Protein

It is recommended that the protein be aliquoted for optimal storage. It is recommended that sterile water (400 μL) be added to the vial to prepare a stock solution of 0.25 mg/mL. Concentration is measured by UV.

The nonstructural protein 3 (nsp3) is one of the SARS-Coronavirus replicase cleaving products, encoded by ORF1a. Nsp3 is thought to be part of the viral replication complex, which is associated with intracellular membranes. Nsp3 contains a papain-like cysteine proteinase and ADP-ribose-1'-monophosphatase activities.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P0DTD1
For Use With (Application) Control, Neutralization
Format Lyophilized
Formulation 20mM tris with 5% mannitol, 500mM NaCl, 5% trehalose, 0.01% Tween 80 and no preservative; pH 7.4
Gene ID (Entrez) 43740578
Molecular Weight (g/mol) 19.87 kDa
Name SARS-CoV-2 NSP3 His-tag
pH Range 7.4
Purification Method Purified
Quantity 100 μg
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Endotoxin Concentration <1.0 EU/μg
Common Name SARS-CoV NSP3
Gene Symbol orf1ab
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Protein Tag His-tag
Sequence SARS-CoV-2 NSP3, amino acids Ile205-Lys379 (Accession # YP_009725299.1) with an N-terminal His-tag
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥95% as determined by SDS-PAGE.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.