Learn More
Invitrogen™ SARS-CoV-2 NSP3 (strain Wuhan-Hu-1) Polyclonal Antibody

Description
Store as concentrated solution. Centrifuge briefly prior to opening vial.
Specifications
Specifications
| Antigen | SARS-CoV-2 NSP3 (strain Wuhan-Hu-1) |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1.01 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 20% glycerol and 0.025% Proclin 300; pH 7 |
| Gene Alias | CoV-2; non-structural protein 3; NSP3 |
| Host Species | Rabbit |
| Immunogen | Recombinant protein encompassing a sequence within the center region of SARS-CoV-2 (COVID-19) nsp3c (Strain:Wuhan-Hu-1). The exact sequence is proprietary. |
| Purification Method | Protein A |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.