Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SARS-CoV-2 Nucleocapsid Recombinant Rabbit Monoclonal Antibody (4Z6T2), Invitrogen™

Catalog No. p-7215294
Encompass
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA536287 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA536287 Supplier Thermo Scientific Supplier No. MA536287

Rabbit Recombinant Monoclonal Antibody

Endotoxin level <1.0 EU/μg of the protein by LAL method. MA5-36286 has been used in ELISA as a detection antibody with MA5-36284 as a capture antibody.

SARS-CoV-2 Nucleocapsid Protein is associated with nucleic acid. It is the most abundant protein for coronavirus. Because of the strong immunogenicity of coronavirus Nucleocapsid, it is believed that SARS-CoV-2 Nucleocapsid Protein has potential value for the diagnosis of the virus. The SARS-CoV-2 N protein is an abundant RNA-binding protein critical for viral genome packaging, yet the molecular details that involve this process are poorly understood. N protein contains three dynamic disordered regions that house putative transiently-helical binding motifs. The two folded domains interact minimally such that full-length N protein is a flexible and multivalent RNA-binding protein. SARS-CoV-2 causes illnesses ranging from the common cold to severe diseases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SARS-CoV-2 Nucleocapsid
Applications ELISA, Functional Assay, Surface Plasmon Resonance
Classification Recombinant Monoclonal
Clone 4Z6T2
Concentration 1.9 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% Braveds MB-1; pH 7.4
Gene N
Gene Accession No. P0DTC9
Gene Alias COVID19 Nucleocapsid, COVID-19 nucleocapsid, COVID19 Spike nucleocapsid, SARS-CoV-2 N
Gene Symbols N
Host Species Rabbit
Immunogen HK293 derived SARS-COV-2 Nucleocapsid corresponding to amino acids 2-419.
Purification Method Affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 43740575
Target Species Virus
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.