Learn More
AdipoGen SARS-CoV-2 (Spike RBD) IgG Serological ELISA Kit (96 wells)
SDP
Supplier:  AdipoGen AG45B0020KI01
Encompass_Preferred
ELISA Kit. Assay Type: Indirect. Detection Type: Colorimetric. Sample Type: Plasma, Serum. This assay is an indirect ELISA assay (based on Spike (RBD) coated protein) for qualitative measurement of human anti-SARS-CoV-2 IgG in serum and plasma. In this assay, a second plate (antigen non-coated Background Plate) is provided to measure the amount of IgG antibodies non-specifically bound to the well. To measure presence of anti-anti-SARS-CoV-2 human IgG antibodies in serum or plasma, net sample optical densities (Net OD) are calculated by subtracting each sample Background Plate OD from the Spike (SARS-CoV-2) antigen plate (Spike Plate) OD.
Catalog No. 50-205-3328
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.