Learn More
ACROBiosystems ACROBiosystems SARS-CoV-2 S protein RBD (N501Y), Fc Tag (MALS verified)
Supplier:  ACROBiosystems SPDC52531MG
Encompass_Preferred
Recombinant Protein;1MG;SARS-CoV-2 S protein RBD (N501Y), Fc Tag (SPD-C5253) is expressed from human 293 cells (HEK293). It contains AA Arg 319 - Lys 537 (Accession # QHD43416.1 (N501Y)). The N501Y mutation was identified in the SARS-CoV-2 variant (known as B.1.1.7, 20B/501Y.V1 or VOC 202012/01) which emerged in the United Kingdom.;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 51.1 kDa. The protein migrates as 55-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized SARS-CoV-2 S protein RBD (N501Y), Fc Tag (Cat. No. SPD-C5253) at 2 μg/mL (100 μL/well) can bind Human ACE2, His Tag (Cat. No. AC2-H52H8) with a linear range of 39-156 ng/mL (QC tested).;Immobilized Human ACE2, His Tag (Cat. No. AC2-H52H8) at 2 μg/mL (100 μL/well) can bind SARS-CoV-2 S protein RBD (N501Y), Fc Tag (Cat. No. SPD-C5253) with a linear range of 0.1-0.8 ng/mL (Routinely tested).
Catalog No. 50-210-7215
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.