Learn More
ACROBiosystems ACROBiosystems SARS-CoV-2 S1 protein (L18F, T20N, P26S, D138Y, R190S, K417T, E484K, N501Y, D614G, H655Y), Fc Tag
Supplier:  ACROBiosystems S1DC5253100UG
Encompass_Preferred
Recombinant Protein;100UG;SARS-CoV-2 S1 protein, Fc Tag (S1D-C5253) is expressed from human 293 cells (HEK293). It contains AA Val 16 - Arg 685 (Accession # QHD43416.1). The mutations (L18F, T20N, P26S, D138Y, R190S, K417T, E484K, N501Y, D614G, H655Y) were identified in the SARS-CoV-2 variant (known as P.1) which emerged in Brazil. ;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 101.5 kDa. The protein migrates as 125-150 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human ACE2, His Tag (Cat. No. AC2-H52H8) at 2 μg/mL (100 μL/well) can bind SARS-CoV-2 S1 protein, Fc Tag (Cat. No. S1D-C5253) with a linear range of 0.4-6 ng/mL (QC tested).;Immobilized SARS-CoV-2 S1 protein, Fc Tag (Cat. No. S1D-C5253) at 2 μg/mL (100 μL/well) can bind Human ACE2, His Tag (Cat. No. AC2-H52H8) with a linear range of 0.2-0.3 μg/mL (Routinely tested).
Catalog No. 50-210-7210
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.