Learn More
AdipoGen SARS-CoV-2 S1 RBDHisB117
SDP
Supplier:  AdipoGen AG40B0205C050
Encompass_Preferred
Recombinant Protein. Receptor-binding domain RBD of SARS-CoV-2 Spike protein S1 aa 319-541 containing the mutation N501Y is fused at the C-terminus to a His-tag. Source HEK 293 cells. Binds to anti-SARS-CoV-2 Spike RBD antibodies in serum or plasma. Binds to human ACE2. Endotoxin <0.01EU/µg purified protein LAL test.. SARS-CoV-2 shares 79.5% sequence identity with SARS-CoV and is 96.2% identical at the genome level to the bat coronavirus BatCoV RaTG133, suggesting it had originated in bats. The coronaviral genome encodes four major structural proteins the Spike S protein, Nucleocapsid N protein, Membrane/Matrix M protein and the Envelope E protein. The SARS Envelope E protein contains a short palindromic transmembrane helical hairpin that seems to deform lipid bilayers, which may explain its role in viral budding and virion envelope morphogenesis. The SARS Membrane/Matrix M protein is one of the major structural viral proteins.
Catalog No. 50-256-2881
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.