Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam SARS-CoV-2 Spike Glycoprotein Peptide, Synthetic, Blocking, 50 ug

Catalog No. 502704042
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-4042 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-4042 Supplier Abcam Supplier No. AB27306350UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Synthetic blocking peptide suitable for blocking assays.

  • Tested Applications: BL
  • Form: Liquid
  • Storage Buffer: pH 7.2, 0.02% Sodium azide, PBS, 0.1% BSA
  • Storage Conditions: Store at -20°C
  • Shipping Temperature: Blue Ice

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule SARS-CoV-2 Spike Glycoprotein
Substrate Properties Peptide-based Substrate
Shipping Condition Blue Ice
Reactivity HCoV
Concentration 0.2 mg/mL
Buffer PBS, 0.1% BSA, 0.02% sodium azide, pH7.2
Content And Storage Store at -20°C.
Form Liquid
Label or Dye Unconjugated
Cross Reactivity SARS-CoV-2
Formulation PBS, 0.1% BSA, 0.02% Sodium azide, pH 7.2
Quantity 50 μg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.