Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SARS-CoV-2 Spike Human anti-Virus, Unconjugated, Clone: AM001414, Proteintech
SDP

Catalog No. 502256617 Shop All Proteintech Group Inc Products
Encompass
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-225-6617 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-225-6617 Supplier Proteintech Group Inc Supplier No. 91361PTG100UG
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Human Recombinant Antibody

The SARS-CoV-2 Spike antibody from Proteintech is a human recombinant antibody generated with virus of virus SARS-CoV-2 Spike. This antibody recognizes virus antigen. The SARS-CoV-2 Spike antibody has been validated for the following applications: Neutralization, ELISA analysis.

Specifications

Antigen SARS-CoV-2 Spike
Applications Neutralization, ELISA
Classification Monoclonal
Clone AM001414
Conjugate Unconjugated
Host Species Human
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 43740568
Target Species Virus
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.