Learn More
ACROBiosystems SARS-CoV-2 Spike NTD 200ug
Supplier:  ACROBiosystems SPDC82H8200UG
Encompass_Preferred
Biotinylated SARS-CoV-2 Spike NTD Protein HisAvitag (BA.2.86/EPI_ISL_18114953) (SPD-C82H8) is expressed from human 293 cells (HEK293). It contains AA Ser 13 - Leu 303 (Accession # QHD43416.1 (ins16MPLFT19IR21TLPP24-26DelA27SS50LH69delV70delV127FG142DY144delF157SR158GN211delL212IV213GL216FH245NA264D)). The spike mutations are identified on the SARS-CoV-2 Omicron variant (Pango lineage: BA.2.86/Denmark/EPI_ISL_18114953).
Catalog No. 50-253-4155
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.