Learn More
Sino Biological SARS-CoV-2 Spike PAb 20ul (Sino Biological)
Supplier:  Sino Biological 40590T3020
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant SARS-CoV-2 (2019-nCoV) Spike ( Catalog#40590-V08B; YP_009724390.1; Ser686-Pro1213). Spike specific IgG was purified by SARS-CoV-2 (2019-nCoV) Spike affinity chromatography.
Catalog No. 50-237-2973
Due to product restrictions, please to purchase or view availability for this product.
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.