Learn More
Invitrogen™ SARS-CoV-2 Spike Protein (RBD) Chimeric Recombinant Rabbit Monoclonal Antibody (P05DHuRb), Alexa Fluor™ 647, eBioscience™
Description
Description: This antibody has a human Fab with a kappa light chain and Rabbit Fc. It recognizes the RBD domain of SARS CoV2 Spike protein. Applications Tested: This P05DHuRb Alexa Fluor™ 647 conjugate has been tested by flow cytometric analysis of HEK 293 cells transfected with a vector encoding full length spike protein from SARS CoV2. This may be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation:633-647 nm; Emission: 668 nm; Laser: Red Laser Filtration: 0.2 μm post-manufacturing filtered.
Specifications
Specifications
| Antigen | SARS-CoV-2 Spike Protein (RBD) Chimeric |
| Applications | Flow Cytometry |
| Classification | Recombinant Monoclonal |
| Clone | P05DHuRb |
| Concentration | 0.2 mg/mL |
| Conjugate | Alexa Fluor 647 |
| Formulation | PBS, proprietary buffer with 0.09% sodium azide; pH 7.2-7.4 |
| Gene Alias | Coronavirus; COVID; Covid 19; COVID19; COVID-19; SARS; SARS Coronavirus; SARS CoV; SARS CoV2; SARS CoV-2; SARS Spike Protein; SARS-CoV2; SARS-CoV-2 |
| Host Species | Rabbit |
| Immunogen | This antibody was developed using computationally derived variants of SARS-CoV antibody structures and is shown to bind Spike Protein S1 subunit (RBD) of SARS CoV2. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.