Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SARS-CoV-2 Spike Protein (RBD) (B.1.617) Fc Chimera Recombinant Protein

Catalog No. PIRP88483
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIRP88483 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIRP88483 Supplier Invitrogen™ Supplier No. RP88483
Add to Cart
Add to Cart

Recombinant Protein

It is recommended that the protein be aliquoted for optimal storage. The mutations were identified in the SARS-CoV-2 varient known as B.1.617 or B.1.617.1 or B.1.617.3.

SARS-CoV-2 (Severe Acute Respiratory Syndrome Coronavirus 2) also known as 2019-nCoV (2019 Novel Coronavirus) is a virus that causes illnesses ranging from the common cold symptoms to severe effects including death and respiratory distress. The CoV Spike (S) protein plays the most important roles in viral attachment, fusion and entry, and serves as a target for development of antibodies, entry inhibitors and vaccines. Of note is the Spike protein receptor-binding domain (RBD, S-RBD) in SARS-CoV-2 Spike protein which binds strongly to human and bat angiotensin-converting enzyme 2 (ACE2) receptors. SARS-CoV-2 RBD exhibits significantly higher binding affinity to ACE2 receptor than SARS-CoV RBD in comparison. S1 contains a receptor-binding domain (RBD) that can specifically bind to angiotensin-converting enzyme 2 (ACE2), the receptor on target cells.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Control, Neutralization
Format Lyophilized
Formulation PBS with 0.01% mannitol, 5-8% trehalose, 0.01% Tween 80 and no preservative; pH 7.4
Molecular Weight (g/mol) 56.4 kDa
Name SARS-CoV-2 Spike Protein (RBD) (B.1.617) Fc Chimera
pH Range 7.4
Purification Method Purified
Quantity 100 μg
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Endotoxin Concentration <1.0 EU/μg
Gene Alias Coronavirus; COVID; Covid 19; COVID19; COVID-19; SARS; SARS Coronavirus; SARS CoV; SARS CoV2; SARS CoV-2; SARS Spike Protein; SARS-CoV2; SARS-CoV-2
Common Name SARS-CoV-2 Spike Protein RBD
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Protein Tag Fc-tag
Sequence SARS-CoV-2 spike RBD (Accession # YP_009724390.1), amino acids Arg319-Phe541 with L452R, E484Q substitutions and a C-terminal human IgG1 Fc region
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System HEK293 cells
Form Lyophilized
Purity or Quality Grade >95% as determined by SDS-PAGE.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.