Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SARS-CoV-2 Spike Protein RBD Omicron (BA.4/BA.5/BA.2.75) Chimeric Recombinant Human Monoclonal Antibody (D003)
GREENER_CHOICE

Catalog No. MA560600
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA560600 100 μg
MA560601 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA560600 Supplier Invitrogen™ Supplier No. MA560600
Add to Cart
Add to Cart

Human Recombinant Monoclonal Antibody

Specifically recognize the SARS-CoV-2 Omicron BA.4/BA.5/BA.5.2 Spike RBD and ECD (Can block the binding between RBD and ACE2). No cross-reactivity with SARS-CoV-2 wild type, Alpha, Beta, Delta variant. Has Medium cross-reactivity in ELISA with SARS-CoV-2 Omicron BA.2.75 Spike RBD.. Has cross-reactivity in ELISA with: SARS-CoV-2 Omicron (BQ.1.1) Spike S1+S2 trimer Protein; SARS-CoV-2 Omicron (BQ.1.1) Spike RBD Protein; SARS-CoV-2 Omicron (BF.7) Spike S1+S2 trimer Protein; SARS-CoV-2 Omicron (BA.4.6/BF.7) Spike RBD Protein; SARS-CoV-2 Omicron (BA.2.75) Spike RBD Protein; SARS-CoV-2 Omicron (BA.2.75) Spike S1+S2 trimer Protein; SARS-CoV-2 Omicron (BA.2.75.2) Spike RBD Protein; SARS-CoV-2 Omicron (BA.2.75.2) Spike S1+S2 trimer Protein; SARS-CoV-2 Omicron (BA.4/BA.5/BA.5.2) Spike S1+S2 trimer Protein; SARS-CoV-2 Omicron (BA.4/BA.5/BA.5.2) Spike RBD Protein; SARS-CoV-2 Omicron (BA.4.6) Spike S1+S2 trimer Protein; No cross-reactivity in ELISA with: SARS-CoV-2 Omicron (XBB.1.16) Spike S1+S2 trimer Protein; SARS-CoV-2 Omicron (XBB.1.5) Spike RBD Protein; SARS-CoV-2 Omicron (XBB.1.5) Spike S1+S2 trimer Protein; SARS-CoV-2 Omicron (XBB.1) Spike S1+S2 trimer Protein; SARS-CoV-2 Omicron (XBB) Spike RBD Protein; SARS-CoV-2 Omicron (BA.2.3.20) Spike RBD Protein; SARS-CoV-2 Omicron (B.1.1.529) S1+S2 trimer Protein; SARS-CoV-2 Omicron (B.1.1.529) Spike RBD Protein; SARS-CoV-2 Alpha (B.1.1.7) Spike S1+S2 trimer Protein; SARS-CoV-2 Alpha (B.1.1.7) Spike RBD Protein; SARS-CoV-2 Beta (B.1.351)...

The severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2) variant Omicron (B 1 1 529) has attracted great concerns since its identification in South Africa Omicron is the fifth variant of concern (VOC) after Alpha (B 1 1 7), Beta (B 1 351), Gamma (P 1) and Delta (B 1 617 2) Omicron variant harbors >30 substitutions/deletions/insertions in the spike, including 15 substitutions in receptor-binding domain (RBD) [PMID: 35397623].
TRUSTED_SUSTAINABILITY

Specifications

Antigen SARS-CoV-2 Spike Protein RBD Omicron (BA.4/BA.5/BA.2.75) Chimeric
Applications ELISA, Neutralization
Classification Recombinant Monoclonal
Clone D003
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Alias 2019-nCoV Spike; COVID-19 Spike; SARS-CoV-2 Spike
Host Species Human
Immunogen Recombinant SARS-CoV-2 Omicron BA.4/BA.5 Spike ECD Protein.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.