Learn More
Invitrogen™ SARS-CoV-2 Spike Protein (RBD) Recombinant Human Monoclonal Antibody (T01KHu)
Description
This antibody recognizes the RBD domain of SARS CoV2 Spike protein. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. This antibody is not recommended for the Western blot application.
Specifications
Specifications
| Antigen | SARS-CoV-2 Spike Protein (RBD) |
| Applications | ELISA, Flow Cytometry, Neutralization, Luminex, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Clone | T01KHu |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.2-7.4 |
| Gene Alias | Coronavirus; COVID; Covid 19; COVID19; COVID-19; SARS; SARS Coronavirus; SARS CoV; SARS CoV2; SARS CoV-2; SARS Spike Protein; SARS-CoV2; SARS-CoV-2 |
| Host Species | Human |
| Immunogen | This antibody was developed using computationally derived variants of SARS-CoV antibody structures and is shown to bind Spike Protein S1 subunit (RBD) of SARS CoV2. |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.