Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SARS-CoV-2 Spike Protein S1 (mutant) His-tag Recombinant Protein

Catalog No. PIRP88512
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIRP88512 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIRP88512 Supplier Invitrogen™ Supplier No. RP88512
Add to Cart
Add to Cart

Recombinant Protein

It is recommended that the protein be aliquoted for optimal storage. In general, recombinant proteins are provided as lyophilized powder.

SARS-CoV-2 (Severe Acute Respiratory Syndrome Coronavirus 2) also known as 2019-nCoV (2019 Novel Coronavirus) is a virus that causes illnesses ranging from the common cold symptoms to severe effects including death and respiratory distress. SARS-CoV-2 Spike Protein is composed of S1 domain and S2 domain (Spike Protein). The Spike protein is the main surface antigen of the coronavirus. The glycosylated Spike protein (as well as the nucleocapsid protein) can be detected in infected cell culture supernatants with antisera from SARS patients. S1 contains a receptor-binding domain (RBD) that can specifically bind to angiotensin-converting enzyme 2 (ACE2), the receptor on target cells. CoV diversity is reflected in the variable Spike proteins (S proteins), which have evolved into forms differing in their receptor interactions and their response to various environmental triggers of virus-cell membrane fusion.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Control, ELISA Standard, Binding Assay
Format Lyophilized
Formulation PBS with 5% mannitol, 5% trehalose and no preservative; pH 7.4
Molecular Weight (g/mol) 76.13 kDa
Name SARS-CoV-2 Spike Protein S1 (mutant) His-tag
pH Range 7.4
Purification Method Purified
Quantity 100 μg
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Endotoxin Concentration <1.0 EU/μg
Gene Alias Coronavirus; COVID; Covid 19; COVID19; COVID-19; SARS; SARS Coronavirus; SARS CoV; SARS CoV2; SARS CoV-2; SARS Spike Protein; SARS-CoV2; SARS-CoV-2
Common Name SARS-CoV-2 Spike Protein S1
Biological Activity Immobilized ACE2 Protein, Human, Recombinant (mFc Tag) (at 2 μg/mL (100 μL/well) can bind SARS-CoV-2 Spike S1 (H49Y, YH145-146 deletion, E484K, D614G) Protein (His Tag), the EC50 of SARS-CoV-2 Spike S1 (H49Y, YH145-146 deletion, E484K, D614G) Protein (His Tag) is 120-600 ng/mL)
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Protein Tag His-tag
Sequence Met1-Arg685
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Activity Immobilized ACE2 Protein, Human, Recombinant (mFc Tag) at 2 μg/mL (100 μL/well) can bind SARS-CoV-2 Spike S1 (H49Y, YH145-146 deletion, E484K, D614G) Protein (His Tag), the EC50 of SARS-CoV-2 Spike S1 (H49Y, YH145-146 deletion, E484K, D614G) Protein (His Tag) is 120-600 ng/mL.
Expression System HEK293 cells
Form Lyophilized
Purity or Quality Grade ≥90% as determined by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.