Learn More
ACROBiosystems SARS-CoV-2 Spike RBD 100ug
Supplier:  ACROBiosystems SPDC5247100UG
Encompass_Preferred
SARS-CoV-2 Spike RBD Protein His Tag (BA.2.86/V483del/USA) (SPD-C5247) is expressed from human 293 cells (HEK293). It contains AA Arg 319 - Lys 537 (Accession # QHD43416.1 (I332VG339HK356TS371FS373PS375FT376AR403KD405NR408SK417NN440KV445HG446SN450DL452WN460KS477NT478KN481KV483delE484KF486PQ498RN501YY505H)). The spike mutations are identified on the SARS-CoV-2 Omicron variant (Pango lineage: BA.2.86/V483del/USA).
Catalog No. 50-253-4161
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.