Learn More
ACROBiosystems SARS-CoV-2 Spike RBD 100ug
Supplier:  ACROBiosystems SRDC52H3100UG
Encompass_Preferred
SARS-CoV-2 S protein RBD (E484K) His Tag (SRD-C52H3) is expressed from human 293 cells (HEK293). It contains AA Arg 319 - Lys 537 (Accession # QHD43416.1 (E484K)). E484K mutaion was identified in the SARS-CoV-2 Beta variant (Pango lineage: B.1.351; other names: 20H/501Y.V2).
Catalog No. 50-253-4106
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.