Learn More
ACROBiosystems Biotinylated SARS-CoV-2 Spike RBD, His,Avitag (BA.4&BA.5/Omicron) (SPD-C82Ew) is expressed from human 293 cells (HEK293). It contains AA Arg 319 - Lys 537 (Accession # QHD43416.1 (G339D, S371F, S373P, S375F, T376A, D405N, R408S, K417N, N440
Supplier:  ACROBiosystems SPDC82EW25UG
Encompass_Preferred
It's been reported that Coronavirus can infect the human respiratory epithelial cells through interaction with the human ACE2 receptor. The spike protein is a large type I transmembrane protein containing two subunits, S1 and S2. S1 mainly contains a receptor binding domain (RBD), which is responsible for recognizing the cell surface receptor. S2 contains basic elements needed for the membrane fusion.The S protein plays key parts in the induction of neutralizing-antibody and T-cell responses, as well as protective immunity.
Catalog No. 50-225-3657