Learn More
Sino Biological SARS-CoV-2 (2019-nCoV) Spike S1-AVI & His Recombinant Protein, Biotinylated 20ug
Supplier:  Sino Biological 40591V27HB20
Encompass_Preferred
A DNA sequence encoding the SARS-CoV-2 (2019-nCoV) Spike S1-AVI & His Recombinant Protein (YP_009724390.1) (Val16-Arg685) was expressed with a c-terminal polyhistidine tagged AVI tag at the C-terminus. The purified protein was biotinylated in vitro.
Catalog No. 50-211-5535
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.