Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SARS Envelope Protein Polyclonal Antibody
GREENER_CHOICE

Catalog No. 01675735
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-675-735 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-675-735 Supplier Invitrogen™ Supplier No. PA141158
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The immunogen is 100% (10/12) homologous to SARS-COV-2 (COVID-19) Envelop protein. The cross reactivity is not empirically tested. Suggested positive control: recombinant fusion protein. Immunogen sequence: YSRVKNLNSS EG.

A novel coronavirus has recently been identified as the causative agent of SARS (Severe Acute Respiratory Syndrome). Coronaviruses are a major cause of upper respiratory diseases in humans. The genomes of these viruses are positive-stranded RNA approximately 27-31kb in length. SARS infection can be mediated by the binding of the viral spike protein, a glycosylated 139 kDa protein and the major surface antigen of the virus, to the angiotensin-converting enzyme 2 (ACE2) on target cells. This binding can be blocked by a soluble form of ACE2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SARS Envelope Protein
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene Alias Coronavirus; COVID; COVID 19; COVID19; COVID-19; SARS; SARS Coronavirus; SARS CoV; SARS CoV2; SARS CoV-2; SARS-CoV2; SARS-CoV-2; SARS-E
Host Species Rabbit
Immunogen Synthetic peptides corresponding to amino acids 59-70 of the putative SARS envelope protein.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.