Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ SARS Spike Protein Antibody Blocking Peptide
SDP

Catalog No. NB398805 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB398805 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB398805 Supplier Novus Biologicals™ Supplier No. NB10056047PEP
Add to Cart
Add to Cart

Specifications

Species Virus
Concentration 1 mg/mL
Content And Storage Store at -80°C. Avoid freeze/thaw cycles.
For Use With (Application) This Peptide is Useful as a Blocking Peptide for NB100-56047
Gene Alias E2, E2 glycoprotein, Human coronavirus spike glycoprotein, Peplomer protein, S, S glycoprotein, SARS coronavirus S protein, SARS coronavirus Spike Protein, SARS CoV, SARS CoV S glycoprotein, SARS CoV Spike protein, SARSCoV, SARS-CoV S protein, SARS-COV Spike, Severe acute respiratory syndrome spike glycoprotein, Severe acute respiratory syndrome virus spike glycoprotein, Spike glycoprotein, VGL2
Quantity 0.1 mg
Research Category Core ESC Like Genes, Immunology, Stem Cell Markers, Virology Bacteria and Parasites
Storage Buffer Peptide dissolved in dH20. Contains no BSA.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.