Learn More
Sino Biological SARS-CoV-2 (BA.1.1) Spike RBD Protein (His Tag) 100ug
Supplier:  Sino Biological 40592V08H129100
Encompass_Preferred
A DNA sequence encoding the SARS-CoV-2 (BA.1.1) Spike RBD (YP_009724390.1, with mutations G339D, R346K, S371L, S373P, S375F, S477N, T478K, E484A, Q493R, G496S, Q498R, N501Y, Y505H)(Arg319-Phe541) was expressed with a polyhistidine tag at the C-terminus. The mutations were identified in the SARS-CoV-2 variant (known as variant BA.1.1).
Catalog No. 50-224-6497
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.