Learn More
Sino Biological Protein SARS-CoV-2 NP-CoV 100u
Supplier:  Sino Biological 40588V07E37100
Encompass_Preferred
A DNA sequence encoding the SARS-CoV-2 (BA 4) Nucleocapsid (YP 009724397 2 with mutation P13L ERS31-33del P151S R203K G204R S413R) (Met1-Ala419) was expressed with a polyhistidine tag at the N-terminus The mutations were identified in the SARS-CoV-2 variant (known as variant BA 4)
Catalog No. 50-224-6470
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.