Learn More
Sino Biological Protein SARS-CoV-2 Spike 100ug
Supplier:  Sino Biological 40589V08B33100
Encompass_Preferred
A DNA sequence encoding the SARS-CoV-2 Spike S1 S2 (YP 009724390 1 with mutations A67V HV69-70del T95I G142D VYY143-145del N211del L212I ins214EPE G339D S371L S373P S375F K417N N440K G446S S477N T478K E484A Q493R G496S Q498R N501Y Y505H T547K D614G H655Y N679K P681H N764K D796Y N856K Q954H N969K L981F) (Met1-Pro1213) was expressed with a polyhistidine tag at the C-terminus The mutations were identified in the SARS-CoV-2 variant (known as variant B 1 1 529) which emerged in the South Africa
Catalog No. 50-222-1641
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.