Learn More
Sino Biological SARS-CoV-2 Spike Antibody (Biotin), Rabbit PAb, Antigen Affinity Purified 100ug
Supplier:  Sino Biological 40591T62B100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant SARS-CoV-2 (2019-nCoV) Spike S1-His Recombiant Protein (HPLC-verified) (Catalog#40591-V08H; YP_009724390.1; Val16-Arg685) and then biotinylated. The specific IgG was purified by SARS-CoV-2 / 2019-nCoV Spike affinity chromatography and then biotinylated.
Catalog No. 50-224-7379
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.