Learn More
Sino Biological Protein SARS-CoV-2 Spike 100ug
Supplier:  Sino Biological 40592V08H103100
Encompass_Preferred
A DNA sequence encoding the SARS-CoV-2 Spike RBD (YP 009724390 1 with mutation E484A) (Arg319-Phe541) was expressed with a polyhistidine tag at the C-terminus
Catalog No. 50-211-4310
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.