Learn More
Sino Biological Protein SARS-CoV-2 Spike 20ug
Supplier:  Sino Biological 40591V49H4B20
Encompass_Preferred
A DNA sequence encoding the SARS-CoV-2 Spike S1 (YP 009724390 1 with mutations (HV69-70 Y145)deletion N501Y A570D D614G P681H) (Met1-Arg685) was expressed with a C-terminal polyhistidine tag followed by an AVI tag The purified protein was biotinylated in vitro The mutations were identified in the SARS-CoV-2 variant (known as variant B 1 1 7) which emerged in the UK
Catalog No. 50-224-7129
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.