Learn More
Sino Biological Protein SARS-CoV-2 Spike 100ug
Supplier:  Sino Biological 40589V08B26100
Encompass_Preferred
A DNA sequence encoding the SARS-CoV-2 Spike S1 S2 (YP 009724390 1 with mutations T19R V70F E156G 157-158 deletion A222V K417N L452R T478K D614G P681R D950N) (Met1-Pro1213) was expressed with a polyhistidine tag at the C-terminus The mutations were identified in the SARS-CoV-2 variant (known as variant AY 2) which emerged in the India
Catalog No. 50-222-1638
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.