Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sartorius Sartolab RF 50

Catalog No. 14558724 Shop All Sartorius Products
Change view
Click to view available options
Pore Size:
0.45 μm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Pore Size
14-558-724 0.45 μm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-558-724 Supplier Sartorius Supplier No. 180F01E8
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sartolab RF 50, PES Vacuum Filtration Units are designed for the sterile filtration of samples up to 50 mL.

  • Highest flow rates with a large filtration surface
  • No loss of protein with a low protein affinity membrane
  • Low hold-up volume due to an optimized membrane support
  • Minimized risk of contamination with a complete ready-to-use unit
  • Can be used as a stand-alone system or with the Sartolab™ Multistation
TRUSTED_SUSTAINABILITY

Specifications

Disposable Single-use
Filter Type Vacuum Filter
Material Polystyrene
Material (Membrane) PES
Pore Size 0.45 μm
Sterility Non-sterile
Volume (Metric) 50 mL
For Use With (Equipment) MultiStation
Sample Volume 50 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.