Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sartorius Sartolab™ Single-Use Polystyrene Storage Bottle
GREENER_CHOICE

Catalog No. 14578813 Shop All Sartorius Products
Change view
Click to view available options
Sample Volume:
150 mL
250 mL
500 mL
1000 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Volume
14-578-813 1000 mL
14-578-811 150 mL
14-578-810 250 mL
14-578-812 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 14-578-813 Supplier Sartorius Supplier No. 180252
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sartorius Sartolab single-use polystyrene bottles for storage of media, buffers, and other aqueous solutions.

  • The bottles can be used with all Sartolab™ RF | BT vacuum filtration units with a 45 mm neck size
TRUSTED_SUSTAINABILITY

Specifications

Disposable Single-use
Material Polystyrene
Material (Membrane) Polystyrene
Sterility Sterile
For Use With (Equipment) Sartolab
Sample Volume 1000 mL
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.