Learn More
Sartorius Accessories for Biosart™ 100 Membrane Filtration
Catalog No. 5090117872 Shop All Sartorius Products
Change view
Click to view available options
Type:
Adapter
Supports
Material:
ABS Copolymer
Polypropylene
Silicone
Stainless Steel
For Use With (Application):
56 mm Supports
Fits Biosart 100 Monitor onto 16840 or 16841 base
Fits Biosart 100 Monitors onto Combisart manifolds 16844, 16842 and 16843
For easy transfer of the membrane onto agar
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Material For Use With (Application)
5090117872 Supports Polypropylene 56 mm Supports
14-555-780 Adapter ABS Copolymer For easy transfer of the membrane onto agar
14-555-778 Adapter Silicone Fits Biosart 100 Monitor onto 16840 or 16841 base
14-555-781 Adapter Stainless Steel Fits Biosart 100 Monitors onto Combisart manifolds 16844, 16842 and 16843
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 5090117872 Supplier Sartorius Supplier No. 16416
Only null left
Add to Cart
Add to Cart
Biosart™ 100 Membrane Filtration is designed specifically for microbiological testing of pharmaceuticals, food, beverages, water and other liquids.
Accessories Descriptions
Adapter, ABS copolymer - 14-555-780
  • Biosart 100 Membrane Lifter
  • For easy transfer of the membrane onto agar
Adapter, Polypropylene - 14-555-779
  • Biosart 100 Adapter
  • Fits Biosart 100 Monitor onto 50 mm supports
Adapter, Silicone - 14-555-778
  • Biosart 100 Adapter
  • 50 mm stainless-steel frit and silicone gasket
  • Fits Biosart 100 Monitor onto 16840 or 16841 base
Adapter, Stainless Steel - 14-555-781
  • Biosart 100 Adapter to accommodate various funnel types
  • Fits Biosart 100 Monitors onto Combisart™ manifolds 16844, 16842 and 16843
Stopper, Silicone - 14-555-817
  • Replacement stopper for 16385
  • 16385 adapts Biosart 100 Monitors on Combisart manifold
TRUSTED_SUSTAINABILITY
Specifications
Disposable Reusable
For Use With (Equipment) Sartorius Biosart 100
Material Polypropylene
Sterility Non-sterile
For Use With (Application) 56 mm Supports
Type Supports