Learn More
Sartorius Accessories for Combisart™ Systems: Forceps
Catalog No. 14555897 Shop All Sartorius Products
Change view
Click to view available options
Display Type:
None
Catalog No. Display Type
14-555-897 None
1 options
Catalog No. 14-555-897 Supplier Sartorius Supplier No. 16625
Add to Cart
Add to Cart
Forceps for use with Sartoris Combisart Systems
  • Stainless Steel
  • Blunt tip
  • Can be flamed, also autoclavable
TRUSTED_SUSTAINABILITY
Specifications
Product Type Tweezers
Display Type None
Material Stainless Steel
For Use With (Application) Firm hold of the membrane filter
For Use With (Equipment) Vacuum filter holders and manifold system
Includes Blunt-edged Tips