Learn More
Sartorius Accessories for Combisart™ Systems: Woulff's Bottle
Catalog No. 14555893 Shop All Sartorius Products
Change view
Click to view available options
Display Type:
None
Catalog No. Display Type
14-555-893 None
1 options
Catalog No. 14-555-893 Supplier Sartorius Supplier No. 16610
Add to Cart
Add to Cart
Bottle for use with Sartorius Combisart System
  • Volume, 500mL
  • With stopcock
  • For protection of vacuum pump
  • Used between suction flask and vacuum source
TRUSTED_SUSTAINABILITY
Specifications
Product Type Woulff's Bottle
Display Type None
Capacity (Metric) 500mL
Material Glass
For Use With (Application) Protection of vacuum pump, used between suction flask and vacuum source
For Use With (Equipment) Used between suction flask and vacuum source
Includes Stop Cock