Learn More
Sartorius Analytical Draft Shield for Quintix™ and Practum™ Toploader Balances
GREENER_CHOICE
Catalog No. 14558110 Shop All Sartorius Products
Change view
Click to view available options
For Use With (Equipment):
Quintix or Practum Balance
Catalog No. For Use With (Equipment)
14-558-110 Quintix or Practum Balance
1 options
Catalog No. 14-558-110 Supplier Sartorius Supplier No. YDS01SQP
Add to Cart
Add to Cart
Analytical draft ring.
Applicable for Quintix or Practum balances with a readability of 10mg and a weighing range of up to 2100g or Quintix/Practum balances with a readability of 0.1g or 1g.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Quintix or Practum Balance
Type Draft Ring