Learn More
Sartorius arium™ Comfort Conbined Water System Accessory, Cartridge
Catalog No. 14556252 Shop All Sartorius Products
Change view
Click to view available options
For Use With (Equipment):
Arium Comfort I and Comfort II
Catalog No. For Use With (Equipment)
14-556-252 Arium Comfort I and Comfort II
1 options
Catalog No. 14-556-252 Supplier Sartorius Supplier No. H2OCPACK
Add to Cart
Add to Cart
Replacement deionization cartridge
  • Cartridge can be used with either arium Comfort I or II systems
TRUSTED_SUSTAINABILITY
Specifications
Case Material High Grade Polypropylene
Certifications/Compliance ASTM type 1
For Use With (Equipment) Arium Comfort I and Comfort II
Type Cartridge Set
WARNING: Cancer - www.P65Warnings.ca.gov