Learn More
Sartorius arium™ Conductivity Meter for Deionization Cartridges
Catalog No. 14556258 Shop All Sartorius Products
Change view
Click to view available options
Display:
Digital
LED
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Display
14-556-258 LED
14-556-257 Digital
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-556-258 Supplier Sartorius Supplier No. 615ALC1
The arium™ line also includes conductivity meters for checking the quality of product water. These convenient options are available in a choice of LED controlled display or digitally controlled display.
The LED controlled conductivity meter enables the user to monitor the conductivity of the product water with a red (>20μS/cm) or green (>20μS/cm) light. If the water does not meet the user's quality requirements, the LED will light up in red to alert the user.
The digitially controlled conductivity meter lets the user monitor the conductivity by individually defined setpoint limits (0.1–200μS/cm). The conductivity measurement range is between 0.056 and 200μS/cm.
TRUSTED_SUSTAINABILITY
Specifications
Type Conductivity Meter
Display LED
For Use With (Application) Checking the quality of product water