Learn More
Sartorius arium™ Foot Switch
Catalog No. 14556267 Shop All Sartorius Products
Change view
Click to view available options
For Use With (Equipment):
Arium Water Purification System
Catalog No. For Use With (Equipment)
14-556-267 Arium Water Purification System
1 options
Catalog No. 14-556-267 Supplier Sartorius Supplier No. H2OAFS1
Add to Cart
Add to Cart
More Convenient Pure Water Dispensing
For external control of the time / volume dispense port for arium 611 systems.
  • Water dispensing at a press of the foot
  • Ideally suited for clean rooms, minimize the risk of contamination
  • Comfortable fatigue-free switching thanks to low height
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Arium Water Purification System
Type Foot Switch