Learn More
Sartorius arium™ Tank Level Switch
Catalog No. 14556260 Shop All Sartorius Products
Change view
Click to view available options
For Use With (Equipment):
Water System
Catalog No. For Use With (Equipment)
14-556-260 Water System
1 options
Catalog No. 14-556-260 Supplier Sartorius Supplier No. H2OALS1
Add to Cart
Add to Cart
For external control of an open gravity tank for arium™ pro systems. The arium™ Tank Level Switch is a system accessory to assist with daily use of arium™ lab water systems.
The level sensor makes it easy to connect an external water storage tank and subsequently fill a tank with ultrapure water.
  • Used with the arium™ multifunction box
  • Flexible water transport to anywhere
  • Can fill every tank system
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Water System
Type Level Sensor
Length (Metric) 3 m Cable
Diameter (Metric) 2.03 cm (bore diameter: 1.65 cm)