Learn More
Sartorius arium™ RO-Module Cleaning Set
Catalog No. 14556251 Shop All Sartorius Products
Change view
Click to view available options
For Use With (Equipment):
Water System
Catalog No. For Use With (Equipment)
14-556-251 Water System
1 options
Catalog No. 14-556-251 Supplier Sartorius Supplier No. H2OCCS
Add to Cart
Add to Cart
Includes
  • Alkaline cleaner HEDTA, ethanolamine, triethanolamine
  • Acidic cleaner HEDTA, phosphoric acid, citric acid
Effective removal of scaling and metal fouling
  • Effective removal of scaling and metal fouling
  • Elimination of organic compounds
  • Dispersion of colloids
  • Stable pH values
  • Gentle on materials
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Water System
Type Arium™ RO-Module Cleaning Set
Includes Alkaline cleaner HEDTA, Ethanolamine, Triethanolamine, Acidic cleaner HEDTA, Phosphoric Acid, Citric Acid
WARNING: Cancer - www.P65Warnings.ca.gov