Learn More
Sartorius arium™ UV Lamp
Catalog No. 14555502 Shop All Sartorius Products
Change view
Click to view available options
For Use With (Equipment):
Arium Water Purification System
Catalog No. For Use With (Equipment)
14-555-502 Arium Water Purification System
1 options
Catalog No. 14-555-502 Supplier Sartorius Supplier No. 611CEL1
Add to Cart
Edge
Add to Cart
Specially designed for arium™ pro 611UV and 611VF laboratory water purifications systems
  • Dual UV radiation at 254 nm and 184 nm wavelengths
  • Used as an bactericide or organic compound breakdown tool
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Arium Water Purification System
Type Lamp