Learn More
Sartorius Arium™ Water Guard
Catalog No. 14555522 Shop All Sartorius Products
Change view
Click to view available options
For Use With (Equipment):
Arium™ Systems
Catalog No. For Use With (Equipment)
14-555-522 Arium™ Systems
1 options
Catalog No. 14-555-522 Supplier Sartorius Supplier No. 610AWG1
Add to Cart
Add to Cart
Arium™ Water Guard detects water leaks early. That means your lab gets optimal protection against water damage. Thanks to its high-grade materials and its finely-tuned sensors, it is the ideal partner for laboratory-grade and ultrapure water systems.
Arium™ Water Guard detects water leaks early. That means your lab gets optimal protection against water damage. Thanks to its high-grade materials and its finely-tuned sensors, Arium™ Water Guard is the ideal partner for laboratory-grade and ultrapure water systems. Leaks are registered by an optical sensor. This automatically closes the inlet line and triggers an acoustic warning signal. In addition, the user can also constantly monitor the system status on the built-in LED display.
  • Reliably protects against leaks
  • Highly sensitive, also suitable for ultrapure water
  • High-grade material, non-corrosive
  • Optic and acoustic alarm
  • Easy to install
  • Integrated wall holder for solenoid valve
  • Automatically blocks the feed water line when a leak is detected
  • Electrically powered
  • Versatile
  • Areas of application: For installation in feed water inlets on Arium™ systems
TRUSTED_SUSTAINABILITY
Specifications
Type Arium Water Guard
For Use With (Application) For installation in feed water inlets on Arium™ systems
Temperature (English) Operating 41°F to 86°F
Temperature (Metric) Operating 5°C to 30°C
For Use With (Equipment) Arium™ Systems
WARNING: Cancer - www.P65Warnings.ca.gov