Learn More
Sartorius Checkweighing Display for Sartorius Balances and Scales
Catalog No. 14557531 Shop All Sartorius Products
Change view
Click to view available options
Three segment: red, green, red
Catalog No.
14-557-531 Three segment: red, green, red
1 options
Catalog No. 14-557-531 Supplier Sartorius Supplier No. YRD11Z
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
For use with CPA, Extend ED, M-Power, Master Pro LA Balances, SE and ME Ultramicro and Microbalances.
For over/under checkweighing
  • 3-segment control display, red – green – red
  • Fits RS232C and checkweighing software
  • 25-pin T-connector
  • 9 ft.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) RS232C and 9 ft. Check Weighing Software
Type Display Indicator