Learn More
Sartorius Arium™ Comfort Pre-Filter
GREENER_CHOICE
Catalog No. 14557603 Shop All Sartorius Products
Change view
Click to view available options
For Use With (Equipment):
Arium™ Comfort I and II
Catalog No. For Use With (Equipment)
14-557-603 Arium™ Comfort I and II
1 options
Catalog No. 14-557-603 Supplier Sartorius Supplier No. H2OCPFCO1
Add to Cart
Add to Cart
Arium™ Comfort Pre-Filter for Arium™ comfort water systems.
  • Pre-filter consisting of high efficient activated carbon combined with a process optimizing catalyst
  • Removes reliable oxidizing agents, heavy metals and particular impurities
  • For intended use with: Arium™ Comfort I Arium™ Comfort II
  • Qty. 1 Unit/Pack
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Arium™ Comfort I and II
Type Pre-treatment Cartridge
WARNING: Cancer - www.P65Warnings.ca.gov