Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sartorius Diafiltration Cups for Vivaspin™ 20

Catalog No. 14558534 Shop All Sartorius Products
Change view
Click to view available options
Quantity:
12/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-558-534 12/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-558-534 Supplier Sartorius Supplier No. VSA005
Add to Cart
Add to Cart

Use with Vivaspin 20

Use with Vivaspin 20
TRUSTED_SUSTAINABILITY

Specifications

Disposable Reusable
Filter Type Ultrafiltration
Sterility Non-sterile
Quantity 12/Pk.
Type Diafiltration Cup
For Use With (Equipment) Vivaspin 20
Sample Volume Up to 2 mL
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.