Learn More
Sartorius M-Power™ Analytical Balances
Catalog No. 14557099 Shop All Sartorius Products
Change view
Click to view available options
Model:
AZ124
AZ214
AZ64
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model
14-557-099 AZ214
14-557-097 AZ64
14-557-098 AZ124
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 14-557-099 Supplier Sartorius Supplier No. AZ214
The balance for first-time users at vocational and professional schools and universities
The innovative weigh cell technology, the rugged construction of the balance housing, the sleek stainless steel weighing pan and the keypad sealed by a membrane overlay assure high dependability and accuracy, even during frequent use. Talent series balances offer various application programs as standard features to make routine work easy: weighing in percent, net-total formulation, weigh averaging / dynamic weighing, counting of small parts and mass unit conversion by toggling between two weight units. Each model comes standard with a bidirectional RS-232C interface port. This means no extra cost if you need to log the balance-generated results on an optional printer or connect a remote display for use in the educational sector.
  • High-contrast LCD
  • All-glass draft shield chamber
  • Bidirectional RS-232C interface port
  • Small Footprint Space Saver
  • Built-in overload protection
  • 4 digital filter levels to adapt the balance to the conditions at the place of use
TRUSTED_SUSTAINABILITY
Specifications
Readability 0.1 mg
Material Durable ABS Housing
Model AZ214
Stabilization Time 3 sec.
Type Analytical Balance