Learn More
Sartorius Moisture Analyzer Software
Catalog No. 14557641 Shop All Sartorius Products
Change view
Click to view available options
Data collection
Catalog No.
14-557-641 Data collection
1 options
Catalog No. 14-557-641 Supplier Sartorius Supplier No. YMW02MA
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
For use with moisture analyzer models MA100, MA150.
For data collection and programming drying functions.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) MA100, MA150 Moisture Analyzer
For Use With (Application) Data collection and programming drying functions
Includes Interface cable, 9-25 pin
Type Software