Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sartorius Phase Separator Papers for separation of Aqueous and Organic Phases using Grade 480

Catalog No. 14555017 Shop All Sartorius Products
Change view
Click to view available options
Diameter (Metric):
12.5 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric)
14-555-017 12.5 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-555-017 Supplier Sartorius Supplier No. FT3602125
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Grade 480 is impregnated with stabilized silicon, thus rendering it hydrophobic: It retains water, but allows solvents to flow through. The flow stops automatically when the entire solvent has passed through. In many applications, this phase separator paper eliminates the need to use separating funnels.

  • This special paper separates aqueous from organic phases
  • The grade 480 is impregnated with a stabilized silicon, thus rendering it hydrophobic
  • It retains water and allows solvents to flow through
  • In many applications, this phase separator paper eliminates the need to use separating funnels
  • They are available in discs and folded filters
TRUSTED_SUSTAINABILITY

Specifications

Filter Type Ultrafiltration
Material Silicone
Sterility Non-sterile
For Use With (Application) Separation of aqueous and organic phases
Quantity 100 Pack
Type Phase Separating Paper
Diameter (English) 4.92 in.
Diameter (Metric) 12.5 cm
For Use With (Equipment) Labscale TFF System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.