Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sartorius Pump Tubing Set for Vivapure™ AdenoPACK

Catalog No. 14558547 Shop All Sartorius Products
Change view
Click to view available options
Diameter (Metric) Inner:
3.2 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Inner
14-558-547 3.2 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-558-547 Supplier Sartorius Supplier No. VSAVPA001
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Complete kit for fast purification of up to 1 x 1013 adenovirus type 5 particles from 500 ml cell culture. Contains 2 AdenoPACK 100 syringe filter units with Sartobind™ ion exchange membrane plus all buffers and further accessories.

Fast purification of up to 1x1013 viral particles Vivapure AdenoPACK 100 is optimally suited for adenovirus purification from up to 200mL cell culture for in vitro transfection. This flexible kit contains two AdenoPACK 100 units, which can be either used in tandem for the purification of up to 200mL cell culture supernatant or individually for purifying virus from up to 60 ml cell culture. The purification is pursued manually with a syringe optimally attached to a retort stand. However, for even more convenience, protocols are provided for optionally running the virus purification with a peristaltic pump or with an infusion pump, in additional to detailed instructions for a manual operation supplied with the Kit. The accessories needed for the operation with a pump are supplied as individual products.

  • High-speed process: completed in 2 hours
  • High virus recoveries
  • Convenient, easy to use
  • For processing 20-200mL cell supernatants
  • Pump tubing set for Vivapure AdenoPACK 100 Male Luer, Female Luer Female Luer Coupler, Tube Set
TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Not Autoclavable
Diameter (English) Inner 0.13 in.
Diameter (Metric) Inner 3.2 mm
Material PVC
Overall Length (English) 47.2 in.
Overall Length (Metric) 1.2 m
For Use With Vivapure™ AdenoPack™ 100 adenovirus (Ad5) purification and concentration kit
Color Clear
Product Type Pump Tubing Set
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.