Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sartorius Reusable Sterility Testing System: Inlet Needles

Catalog No. 14555845 Shop All Sartorius Products
Change view
Click to view available options
Type:
Long Inlet Needles
Short Inlet Needles
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
14-555-845 Short Inlet Needles
14-555-844 Long Inlet Needles
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-555-845 Supplier Sartorius Supplier No. 169643
Only null left
Add to Cart
Add to Cart

Inlet Needles for use with Reusable Sterility Testing System.

Inlet Needle - 14-555-844

  • Stainless Steel
  • For ampoule testing, infusion bottles
  • Pierces rubber septa on side to prevent blockage by rubber particles from piercing
  • Dia. outlet hose nipple, 5 mm
  • Size, long

Inlet Needle - 14-555-845

  • Stainless Steel
  • For ampoule testing, infusion bottles
  • Pierces rubber septa on side to prevent blockage by rubber particles from piercing
  • Dia. outlet hose nipple, 5 mm
  • Size, short
TRUSTED_SUSTAINABILITY

Specifications

Disposable Reusable
Filter Type Inlet needle
For Use With (Equipment) Reusable Sterility Testing System
Material Stainless Steel
Sterility Non-sterile
Type Short Inlet Needles
Diameter (English) Inside 0.2 in.
Diameter (Metric) Inside 5 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.